missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABPA Antibody (8C1B10), Novus Biologicals™
Mouse Monoclonal Antibody
£161.00
Specifications
| Antigen | GABPA |
|---|---|
| Clone | 8C1B10 |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
GABPA Monoclonal specifically detects GABPA in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| GABPA | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| E4TF1-60, E4TF1ATranscription factor E4TF1-60, GA binding protein transcription factor, alpha subunit 60kDa, GA-binding protein alpha chain, GA-binding protein transcription factor, alpha subunit (60kD), GABP subunit alpha, human nuclear respiratory factor-2 subunit alpha, 10, NFT2, NRF2, NRF2A, nuclear respiratory factor 2 alpha subunit, Nuclear respiratory factor 2 subunit alpha | |
| GABPA | |
| IgG1 | |
| Unpurified | |
| 51 kDa |
| 8C1B10 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| 2551 | |
| Purified recombinant fragment of human GABPA (aa120-190) expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title