missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABPA Antibody (8C1B10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47416-0.025ml
This item is not returnable.
View return policy
Description
GABPA Monoclonal specifically detects GABPA in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| GABPA | |
| Monoclonal | |
| Unconjugated | |
| E4TF1-60, E4TF1ATranscription factor E4TF1-60, GA binding protein transcription factor, alpha subunit 60kDa, GA-binding protein alpha chain, GA-binding protein transcription factor, alpha subunit (60kD), GABP subunit alpha, human nuclear respiratory factor-2 subunit alpha, 10, NFT2, NRF2, NRF2A, nuclear respiratory factor 2 alpha subunit, Nuclear respiratory factor 2 subunit alpha | |
| Mouse | |
| 51 kDa | |
| 0.025 mL | |
| Primary | |
| Human | |
| Ascites |
| Western Blot | |
| 8C1B10 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| GABPA | |
| Purified recombinant fragment of human GABPA (aa120-190) expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| 2551 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction