missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GABARAP Recombinant Rabbit Monoclonal Antibody (23GB5490), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | GABARAP |
|---|---|
| Clone | 23GB5490 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088174
![]() |
Invitrogen™
MA552866 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088496
|
Invitrogen™
MA552867 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GABARAP Recombinant Monoclonal antibody specifically detects GABARAP in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| GABARAP | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O95166, P60517, Q9DCD6 | |
| 11337, 56486, 58974 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5490 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| ATG8A; FLC3B; FLJ25768; GABA type A receptor-associated protein; GABA(A) receptor-associated protein; GABA-A receptor-associated protein; Gabarap; GABARAP a; GABARAP-a; gamma-aminobutyric acid receptor associated protein; gamma-aminobutyric acid receptor-associated protein; gamma-aminobutyric acid reseptor associated protein; HT004; hypothetical protein; MGC120154; MGC120155; MM46 | |
| A synthesized peptide derived from human GABARAP (10-64AA). | |
| Primary | |
| -20°C | |
| GABARAP |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto