missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABARAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99474-100ul
This item is not returnable.
View return policy
Description
GABARAP Polyclonal antibody specifically detects GABARAP in Human samples. It is validated for Western Blot
Specifications
| GABARAP | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| ATG8A, FLC3B, FLJ25768, GABA(A) receptor-associated proteinMGC120154, gamma-aminobutyric acid receptor-associated protein, MM46MGC120155 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human GABARAP. | |
| 100 μg | |
| Autophagy, Golgi Apparatus Markers, Macroautophagy, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 11337 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction