missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABARAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £255.00
Specifications
| Antigen | GABARAP |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18651146
|
Novus Biologicals
NBP2-99474-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18634955
|
Novus Biologicals
NBP2-99474-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GABARAP Polyclonal antibody specifically detects GABARAP in Human samples. It is validated for Western BlotSpecifications
| GABARAP | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Golgi Apparatus Markers, Macroautophagy, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 0.2 um filtered solution in PBS | |
| 11337 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ATG8A, FLC3B, FLJ25768, GABA(A) receptor-associated proteinMGC120154, gamma-aminobutyric acid receptor-associated protein, MM46MGC120155 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human GABARAP. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title