missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-AR alpha 5 Antibody (5C11), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | GABA-AR alpha 5 |
|---|---|
| Clone | 5C11 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043054
|
Novus Biologicals
NBP3-26561-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043835
|
Novus Biologicals
NBP3-26561-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GABA-AR alpha 5 Monoclonal antibody specifically detects GABA-AR alpha 5 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifikationer
| GABA-AR alpha 5 | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| GABA(A) receptor subunit alpha-5, gamma-aminobutyric acid (GABA) A receptor, alpha 5, gamma-aminobutyric acid receptor subunit alpha-5, MGC138184 | |
| A synthesized peptide derived from Human GABA-AR alpha 5 [UniProt P31644] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 5C11 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, Neuronal Cell Markers, Neurotransmission | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 2558 | |
| IgG | |
| Affinity purified |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel