missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-AR alpha 5 Antibody (5C11), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26561-100ul
This item is not returnable.
View return policy
Description
GABA-AR alpha 5 Monoclonal antibody specifically detects GABA-AR alpha 5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| GABA-AR alpha 5 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| 5C11 | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
| GABA(A) receptor subunit alpha-5, gamma-aminobutyric acid (GABA) A receptor, alpha 5, gamma-aminobutyric acid receptor subunit alpha-5, MGC138184 | |
| A synthesized peptide derived from Human GABA-AR alpha 5 [UniProt P31644] | |
| 100 μL | |
| Cellular Markers, Neuronal Cell Markers, Neurotransmission | |
| 2558 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction