missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-AR alpha 1 Antibody (1F4) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
£196.00 - £548.00
Specifications
| Antigen | GABA-AR alpha 1 |
|---|---|
| Clone | 1F4 |
| Concentration | 1 mg/mL |
| Applications | Immunofluorescence, X-Ray Crystallography |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18652637
|
Novus Biologicals
NBP3-09047-0.025mg |
0.025 mg |
£196.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18660807
|
Novus Biologicals
NBP3-09047-0.2mg |
0.2 mg |
£548.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GABA-AR alpha 1 Monoclonal antibody specifically detects GABA-AR alpha 1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, X-Ray CrystallographySpecifications
| GABA-AR alpha 1 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ECA4, EJM, EJM5GABA(A) receptor subunit alpha-1, gamma-aminobutyric acid (GABA) A receptor, alpha 1, gamma-aminobutyric acid receptor subunit alpha-1 | |
| This antibody was raised against alpha 1 beta 2 gamma 2 GABA-A receptor. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1F4 | |
| Immunofluorescence, X-Ray Crystallography | |
| Unconjugated | |
| Rabbit | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| PBS | |
| 2554 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title