missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-AR alpha 1 Antibody (1F4) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-09047-0.2mg
This item is not returnable.
View return policy
Description
GABA-AR alpha 1 Monoclonal antibody specifically detects GABA-AR alpha 1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, X-Ray Crystallography
Specifications
| GABA-AR alpha 1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence, X-Ray Crystallography | |
| 1F4 | |
| Unconjugated | |
| ECA4, EJM, EJM5GABA(A) receptor subunit alpha-1, gamma-aminobutyric acid (GABA) A receptor, alpha 1, gamma-aminobutyric acid receptor subunit alpha-1 | |
| This antibody was raised against alpha 1 beta 2 gamma 2 GABA-A receptor. | |
| 0.2 mg | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 2554 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction