missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G6PC Antibody (CL5818), Novus Biologicals™
Mouse Monoclonal Antibody
£292.00 - £435.00
Specifications
| Antigen | G6PC |
|---|---|
| Clone | CL5818 |
| Dilution | Immunohistochemistry-Paraffin 1:5000 - 1:10000 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18202652
|
Novus Biologicals
NBP2-61433 |
100 μL |
£435.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18688967
|
Novus Biologicals
NBP2-61433-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
G6PC Monoclonal specifically detects G6PC in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| G6PC | |
| Immunohistochemistry-Paraffin 1:5000 - 1:10000 | |
| Unconjugated | |
| Mouse | |
| EC 3.1.3.9, G6Pase, G-6-Pase, G6Pase-alpha, G6PT, glucose-6-phosphatase, Glucose-6-phosphatase alpha, glucose-6-phosphatase, catalytic (glycogen storage disease type I, von Gierkedisease), glucose-6-phosphatase, catalytic subunit, GSD1, GSD1a, MGC163350 | |
| G6PC | |
| IgG2b | |
| Protein A purified |
| CL5818 | |
| Monoclonal | |
| Purified | |
| Human | |
| 2538 | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: DFGIQSTHYLQVNYQDSQD | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title