missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G6PC Antibody (CL5818), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61433-25ul
This item is not returnable.
View return policy
Description
G6PC Monoclonal specifically detects G6PC in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| G6PC | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2), containing 40% glycerol with 0.02% Sodium Azide | |
| G6PC | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: DFGIQSTHYLQVNYQDSQD | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2b |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL5818 | |
| Immunohistochemistry-Paraffin 1:5000 - 1:10000 | |
| EC 3.1.3.9, G6Pase, G-6-Pase, G6Pase-alpha, G6PT, glucose-6-phosphatase, Glucose-6-phosphatase alpha, glucose-6-phosphatase, catalytic (glycogen storage disease type I, von Gierkedisease), glucose-6-phosphatase, catalytic subunit, GSD1, GSD1a, MGC163350 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 2538 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction