missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G6b/C6orf25 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£156.00 - £301.00
Specifications
| Antigen | G6b/C6orf25 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.5-2 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18117661
|
Novus Biologicals
NBP2-24505SS |
0.025 mg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18149971
|
Novus Biologicals
NBP2-24505 |
0.1 mg |
£301.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
G6b/C6orf25 Polyclonal specifically detects G6b/C6orf25 in Human, Primate samples. It is validated for Western Blot.Specifications
| G6b/C6orf25 | |
| Western Blot 0.5-2 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Human, Primate | |
| chromosome 6 open reading frame 25, G6b, immunoglobulin receptor, MGC142279, MGC142281, NG31, protein G6b | |
| MPIG6B | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_b), rhesus monkey, bovine, chicken, horse and pig, 92% homologous in cat, mouse and rat, and 78% homologous in dog. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide | |
| 80739 | |
| A portion of amino acids 1-50 of human G6B was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title