missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G6b/C6orf25 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24505
This item is not returnable.
View return policy
Description
G6b/C6orf25 Polyclonal specifically detects G6b/C6orf25 in Human, Primate samples. It is validated for Western Blot.
Specifications
| G6b/C6orf25 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide | |
| MPIG6B | |
| A portion of amino acids 1-50 of human G6B was used as the immunogen. | |
| 0.1 mg | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_b), rhesus monkey, bovine, chicken, horse and pig, 92% homologous in cat, mouse and rat, and 78% homologous in dog. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 0.5-2 ug/ml | |
| chromosome 6 open reading frame 25, G6b, immunoglobulin receptor, MGC142279, MGC142281, NG31, protein G6b | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 80739 | |
| Human, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction