missing translation for 'onlineSavingsMsg'
Learn More

G3BP2 Mouse, anti-Human, Clone: 2E5G3, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen G3BP2
Clone 2E5G3
Concentration 1000 μg/mL
Applications Western Blot, Immunofluorescence, ELISA
Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18078205
View Documents
Proteintech
68580-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18006215
View Documents
Proteintech
68580-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The G3BP2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human G3BP2. This antibody recognizes human, rat antigen. The G3BP2 antibody has been validated for the following applications: IF, WB, ELISA analysis.

Specifications

Specifications

G3BP2
1000 μg/mL
Liquid
Human, Rat
G3BP 2, G3BP2, KIAA0660
Primary
2E5G3
Western Blot, Immunofluorescence, ELISA
Mouse
9908
IgG1
Store at -20°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.