Learn More
G3BP2 Mouse, anti-Human, Clone: 2E5G3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68580-1-IG-20UL
This item is not returnable.
View return policy
Description
The G3BP2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human G3BP2. This antibody recognizes human, rat antigen. The G3BP2 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| G3BP2 | |
| 2E5G3 | |
| G3BP 2, G3BP2, KIAA0660 | |
| 20 μL | |
| 9908 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, Immunofluorescence, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat | |
| 150 kDa | |
| IgG1 |