Learn More
G3BP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-82976
 View more versions of this product
Product Code. 18700394
  • £452.00 / 0.10mL
Description
G3BP2 Polyclonal antibody specifically detects G3BP2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rabbit
0.1 mL