missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G3BP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | G3BP1 |
|---|---|
| Concentration | 0.25 mg/mL |
| Applications | Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16841084
|
Proteintech
13057-2-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16831084
|
Proteintech
13057-2-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
May be a regulated effector of stress granule assembly. Phosphorylation-dependent sequence-specific endoribonuclease in vitro. Cleaves exclusively between cytosine and adenine and cleaves c-myc mRNA preferentially at the 3′UTR. ATP- and magnesium-dependenSpecifications
| G3BP1 | |
| Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| P97855, Q13283 | |
| 10146, 171092, 27041 | |
| G3BP1 Fusion Protein Ag3728 | |
| Primary | |
| -20°C | |
| G3BP1 |
| 0.25 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| G3BP, G3BP 1, G3BP1, HDH VIII | |
| G3bp1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title