missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
May be a regulated effector of stress granule assembly. Phosphorylation-dependent sequence-specific endoribonuclease in vitro. Cleaves exclusively between cytosine and adenine and cleaves c-myc mRNA preferentially at the 3′UTR. ATP- and magnesium-dependen
Specifications
Specifications
| Antigen | G3BP1 |
| Applications | Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | G3BP1 |
| Gene Accession No. | P97855, Q13283 |
| Gene Alias | G3BP, G3BP 1, G3BP1, HDH VIII |
| Gene Symbols | G3bp1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?