missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G protein alpha inhibitor 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £488.00
Specifications
| Antigen | G protein alpha inhibitor 1 |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Regulatory Status | RUO |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18254505
|
Novus Biologicals
NBP2-59000 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18611077
|
Novus Biologicals
NBP2-59000-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
G protein alpha inhibitor 1 Polyclonal specifically detects G protein alpha inhibitor 1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| G protein alpha inhibitor 1 | |
| Unconjugated | |
| RUO | |
| Human | |
| 2770 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ARQLFVLAGAAEEGFMTAELAGVIKR | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| Adenylate cyclase-inhibiting G alpha protein, Gi, Gi1 protein alpha subunit, guanine nucleotide binding protein (G protein), alpha inhibiting activitypolypeptide 1, guanine nucleotide-binding protein G(i) subunit alpha-1 | |
| GNAI1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title