missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G protein alpha inhibitor 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-59000
This item is not returnable.
View return policy
Description
G protein alpha inhibitor 1 Polyclonal specifically detects G protein alpha inhibitor 1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| G protein alpha inhibitor 1 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Adenylate cyclase-inhibiting G alpha protein, Gi, Gi1 protein alpha subunit, guanine nucleotide binding protein (G protein), alpha inhibiting activitypolypeptide 1, guanine nucleotide-binding protein G(i) subunit alpha-1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| GNAI1 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ARQLFVLAGAAEEGFMTAELAGVIKR | |
| 100 μL | |
| Signal Transduction | |
| 2770 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction