missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G protein alpha 12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-55321
This item is not returnable.
View return policy
Description
G protein alpha 12 Polyclonal specifically detects G protein alpha 12 in Human samples. It is validated for Western Blot.
Specifications
| G protein alpha 12 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| G alpha-12, gep, G-protein subunit alpha-12, guanine nucleotide binding protein (G protein) alpha 12, guanine nucleotide-binding protein subunit alpha-12, MGC104623, MGC99644, NNX3, RMP, WUGSC:H_GS165O14.2 | |
| Rabbit | |
| 44 kDa | |
| 100 μL | |
| Signal Transduction | |
| 2768 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q03113 | |
| GNA12 | |
| Synthetic peptides corresponding to GNA12(guanine nucleotide binding protein (G protein) alpha 12) The peptide sequence was selected from the middle region of GNA12. Peptide sequence TFQLYVPALSALWRDSGIREAFSRRSEFQLGESVKYFLDNLDRIGQLNYF. | |
| Affinity purified | |
| RUO | |
| Primary | |
| This product is specific to Subunit or Isoform: alpha-12. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction