missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ g-linked ATP agarose resin

Product Code. 18721454 Shop All Bio Techne Products
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
This item is not returnable. View return policy

Product Code. 18721454

Brand: Novus Biologicals™ 5100002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Affinity resin for purification of enzymes that bind nucleotides and related molecules

Resins in which ATP is linked via the gamma-phosphate have been valuable in identifying proteins in the purine-binding proteome, which includes kinases, heat shock proteins and other ATP-binding proteins.

  • ATP Agarose resin comprises ATP attached to agarose beads via its gamma-phosphate
  • Two forms of resin are available with low and high ligand substitution
  • Long hydrophilic spacer (14-atom) minimises unwanted hydrophobic interactions and facilitates unhindered interactions with biomolecules

Recommended Storage

Store at 4°C. Do not freeze.

TRUSTED_SUSTAINABILITY

Specifications

Formulation High ligand substitution (8-12μmol/mL)
Content And Storage 4°C. Do not freeze.
Quantity 2 mL
Type Resin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.