missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FYTTD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-74256
This item is not returnable.
View return policy
Description
FYTTD1 Polyclonal specifically detects FYTTD1 in Human samples. It is validated for Western Blot.
Specifications
| FYTTD1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| FYTTD1 | |
| Synthetic peptides corresponding to the middle region of FYTTD1. Immunizing peptide sequence PSQLSRKNNIPANFTRSGNKLNHQKDTRQATFLFRRGLKVQAQLNTEQLL. | |
| Affinity purified | |
| RUO | |
| 84248 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| forty-two-three domain containing 1, Forty-two-three domain-containing protein 1, Protein 40-2-3, UAP56-interacting factor, UIFDKFZp761B1514 | |
| Rabbit | |
| 25 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: . | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering