missing translation for 'onlineSavingsMsg'
Learn More

FXR/NR1H4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications

£186.00 - £412.00

Specifications

Antigen FXR/NR1H4
Concentration 2.4 mg/ml
Dilution Western Blot 1-4 ug/ml, Immunocytochemistry/Immunofluorescence 1:100
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18054974
View Documents
Novus Biologicals
NB400-153SS
0.025 mL
£186.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18799443
View Documents
Novus Biologicals
NB400-153
£412.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FXR/NR1H4 Polyclonal specifically detects FXR/NR1H4 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

FXR/NR1H4
Western Blot 1-4 ug/ml, Immunocytochemistry/Immunofluorescence 1:100
Polyclonal
Rabbit
Cancer, Chromatin Research, Lipid and Metabolism, Signal Transduction
Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide
9971
A synthetic peptide made to the C-terminus of the human FXR protein. [UniProt# Q96RI1]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
2.4 mg/ml
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
Human, Mouse
bile acid receptor, farnesoid X receptor, Farnesoid X-activated receptor, Farnesol receptor HRR-1, FXRBAR, HRR-1, HRR1MGC163445, Nuclear receptor subfamily 1 group H member 4, nuclear receptor subfamily 1, group H, member 4, Retinoid X receptor-interacting protein 14, RIP14RXR-interacting protein 14
NR1H4
IgG
Affinity Purified
66 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.