missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FXR/NR1H4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB400-153SS
This item is not returnable.
View return policy
Description
FXR/NR1H4 Polyclonal specifically detects FXR/NR1H4 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| FXR/NR1H4 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide | |
| NR1H4 | |
| A synthetic peptide made to the C-terminus of the human FXR protein. [UniProt# Q96RI1] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 2.4 mg/ml | |
| Western Blot 1-4 ug/ml, Immunocytochemistry/Immunofluorescence 1:100 | |
| bile acid receptor, farnesoid X receptor, Farnesoid X-activated receptor, Farnesol receptor HRR-1, FXRBAR, HRR-1, HRR1MGC163445, Nuclear receptor subfamily 1 group H member 4, nuclear receptor subfamily 1, group H, member 4, Retinoid X receptor-interacting protein 14, RIP14RXR-interacting protein 14 | |
| Rabbit | |
| 66 kDa | |
| 0.025 mL | |
| Cancer, Chromatin Research, Lipid and Metabolism, Signal Transduction | |
| 9971 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction