missing translation for 'onlineSavingsMsg'
Learn More

Furfuryl Methacrylate (stabilized with MEHQ), TRC

Product Code. 30398743
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398743 100 mg 100mg
30404227 50 mg -
30406203 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30398743 Supplier Toronto Research Chemicals Supplier No. TRCF822530100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3454-28-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.17
IUPAC Name furan-2-ylmethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCc1occc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H10O3/c1-7(2)9(10)12-6-8-4-3-5-11-8/h3-5H,1,6H2,2H3
Formula Weight 166.063
Chemical Name or Material Furfuryl Methacrylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.