CAS RN 3454-28-2
IUPAC Name:
(furan-2-yl)methyl 2-methylprop-2-enoate
Synonyms:
furan-2-ylmethyl 2-methylprop-2-enoate
Molecular Weight (g/mol):
166.18
Molecular Formula:
C9H10O3
InChi Key:
DWXAVNJYFLGAEF-UHFFFAOYSA-N
SMILES:
CC(=C)C(=O)OCC1=CC=CO1
1
–
1
of
1
results
1
Furfuryl Methacrylate (stabilized with MEHQ), TRC
CAS: 3454-28-2 Molecular Formula: C9H10O3 Molecular Weight (g/mol): 166.17 IUPAC Name: furan-2-ylmethyl 2-methylprop-2-enoate SMILES: CC(=C)C(=O)OCc1occc1