missing translation for 'onlineSavingsMsg'
Learn More

Fumaric Acid Monomethyl Ester-d5, TRC

Product Code. 30404462
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30404462 50 mg
30393655 10 mg
30403937 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30404462 Supplier Toronto Research Chemicals Supplier No. TRCF50039350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1616345-45-9
Molecular Formula C5HD5O4
Molecular Weight (g/mol) 135.13
Synonym (2E)-2-Butenedioic Acid 1-Methyl Ester-d5, (E)-2-Butenedioic Acid Monomethyl Ester-d5, Fumaric Acid Methyl Ester-d5, Methyl Hydrogen Fumarate-d5, Monomethyl Fumarate-d5
SMILES OC(/C([2H])=C([2H])/C(OC([2H])([2H])[2H])=O)=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C5H6O4/c1-9-5(8)3-2-4(6)7/h2-3H,1H3,(H,6,7)/b3-2+/i1D3,2D,3D
Chemical Name or Material Fumaric Acid Monomethyl Ester-d5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.