CAS RN 1616345-45-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fumaric Acid Monomethyl Ester-d5, TRC
CAS: 1616345-45-9 Molecular Formula: C5HD5O4 Molecular Weight (g/mol): 135.13 Synonym: (2E)-2-Butenedioic Acid 1-Methyl Ester-d5,(E)-2-Butenedioic Acid Monomethyl Ester-d5,Fumaric Acid Methyl Ester-d5,Methyl Hydrogen Fumarate-d5,Monomethyl Fumarate-d5 SMILES: OC(/C([2H])=C([2H])/C(OC([2H])([2H])[2H])=O)=O