Learn More
Agilent BioTek Full-Size Dichroic Mirror, Ex. 475 to 520, Em. 530 to 670
Product Code. 16502282 Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
Cytation 5, Synergy H1
Unit Size:
Each
Product Code. 16502282
Brand: Agilent BioTek 7137525
For use with Cytation 5, Synergy H1
  • Accessory for Agilent BioTek instruments range of microplate washers, readers and cell imagers
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Cytation 5, Synergy H1