missing translation for 'onlineSavingsMsg'
Learn More

MULTIROIR Full Eurobox Container

Product Code. 17236890
Change view
Click to view available options
Dimensions (L x W x H):
400 x 300 x 117 mm
400 x 300 x 120 mm
400 x 300 x 220 mm
400 x 300 x 290 mm
600 x 400 x 100 mm
600 x 400 x 220 mm
600 x 400 x 320 mm
600 x 400 x 420 mm
Color:
Blue
Gray
Red
White
Unit Size:
Each
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Color unitSize
17236890 400 x 300 x 117 mm Gray Each
17246890 400 x 300 x 220 mm Gray Each
17256890 400 x 300 x 290 mm Gray Each
17266890 600 x 400 x 220 mm Gray Each
17276890 600 x 400 x 320 mm Gray Each
17286890 600 x 400 x 420 mm Gray Each
17296890 400 x 300 x 120 mm Blue Each
17206900 400 x 300 x 120 mm White Each
17216900 400 x 300 x 120 mm Red Each
17226900 400 x 300 x 220 mm Blue Each
17236900 400 x 300 x 220 mm White Each
17246900 400 x 300 x 220 mm Red Each
17256900 600 x 400 x 100 mm Blue Each
17266900 600 x 400 x 100 mm White Each
17276900 600 x 400 x 100 mm Red Each
17286900 600 x 400 x 220 mm Blue Each
17296900 600 x 400 x 220 mm White Each
17206910 600 x 400 x 220 mm Red Each
17216910 600 x 400 x 320 mm Blue Each
17226910 600 x 400 x 320 mm White Each
17236910 600 x 400 x 320 mm Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
This item is not returnable. View return policy
Product Code. 17236890 Supplier MULTIROIR Supplier No. BA12001

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Full Eurobox container with solid bottoms and solid walls, ideal for the storage and transport of goods

  • Full Eurobox container
  • The best quality / price ratio on the market
  • Solid bottoms and solid walls
  • Ideal for the storage and transport of goods
  • Stackable and high load capacity
  • Temperature resistant from -20 ° C to + 110 ° C

Specifications

Type Storage Box
Material Polypropylene
Color Gray
Shape Rectangular
Dimensions (L x W x H) 400 x 300 x 117 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.