missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSCN3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79542
This item is not returnable.
View return policy
Description
FSCN3 Polyclonal specifically detects FSCN3 in Human samples. It is validated for Western Blot, PCR.
Specifications
| FSCN3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| fascin (Strongylocentrotus purpuratus) homolog 3 (actin-bundling protein, fascin homolog 3, actin-bundling protein, testicular (Strongylocentrotuspurpuratus), fascin-3, testicular), Testis fascin | |
| Rabbit | |
| 55 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Dog: 86%; Rat: 86%; Mouse: 86%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot, PCR | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, PCR | |
| NP_065102 | |
| FSCN3 | |
| Synthetic peptide directed towards the middle region of human FSCN3The immunogen for this antibody is FSCN3. Peptide sequence WGKFALNFCIELQGSNLLTVLAPNGFYMRADQSGTLLADSEDITRECIWE. | |
| Affinity purified | |
| RUO | |
| 29999 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction