missing translation for 'onlineSavingsMsg'
Learn More

Fructose-alanine (mixture of diastereomers), TRC

Product Code. 30402291
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402291 5 mg 5mg
30407036 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402291 Supplier Toronto Research Chemicals Supplier No. TRCF7925205MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16124-24-6
Molecular Formula C9H17NO7
Molecular Weight (g/mol) 251.23
Synonym N-(1-Deoxy-D-fructos-1-yl)-L-alanine, (S)-1-[(1-Carboxyethyl)amino]-1-deoxy-D-fructose, 1-Alanino-1-deoxy-D-fructose, 1-Deoxy-1-L-alanine-D-fructose, N-(1-Deoxy-D-fructosyl)-L-alanine
SMILES C[C@H](C(=O)O)NCC1([C@H]([C@@H]([C@@H](CO1)O)O)O)O

Specifications

InChI Formula InChI=1S/C9H17NO7/c1-4(8(14)15)10-3-9(16)7(13)6(12)5(11)2-17-9/h4-7,10-13,16H,2-3H2,1H3,(H,14,15)/t4-,5-,6-,7+,9?/m1/s1
Quantity 5 mg
Formula Weight 251.1005
Percent Purity >95%
Shipping Condition Dry Pack
Chemical Name or Material Fructose-alanine (Mixture of Diastereomers)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.