CAS RN 16124-24-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fructose-alanine (mixture of diastereomers), TRC
CAS: 16124-24-6 Molecular Formula: C9H17NO7 Molecular Weight (g/mol): 251.23 Synonym: N-(1-Deoxy-D-fructos-1-yl)-L-alanine,(S)-1-[(1-Carboxyethyl)amino]-1-deoxy-D-fructose,1-Alanino-1-deoxy-D-fructose,1-Deoxy-1-L-alanine-D-fructose,N-(1-Deoxy-D-fructosyl)-L-alanine SMILES: C[C@H](C(=O)O)NCC1([C@H]([C@@H]([C@@H](CO1)O)O)O)O