missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FRS2 Recombinant Rabbit Monoclonal Antibody (1K7X4)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558041
This item is not returnable.
View return policy
Description
Immunogen sequence: EEQVHTYVNT TGVQEERKNR TSVHVPLEAR VSNAESSTPK EEPSSIEDRD PQILLEPEGV KFVLGPTPVQ KQLMEKEKLE QLGRDQVSGS GANNTEWDTG.
Adapter protein that links activated FGR and NGF receptors to downstream signaling pathways. Plays an important role in the activation of MAP kinases and in the phosphorylation of PIK3R1, the regulatory subunit of phosphatidylinositol 3-kinase, in response to ligand-mediated activation of FGFR1. Modulates signaling via SHC1 by competing for a common binding site on NTRK1. [UniProt].
Specifications
| FRS2 | |
| Recombinant Monoclonal | |
| 1.27 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q8C180, Q8WU20 | |
| FRS2 | |
| A synthetic peptide corresponding to a sequence within amino acids 190-289 of human FRS2 (NP_006645.3). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 1K7X4 | |
| Unconjugated | |
| FRS2 | |
| 4732458E18; C330018A15Rik; FGFR signalling adaptor; FGFR substrate 2; FGFR-signaling adaptor SNT; fibroblast growth factor receptor substrate 2; FRS1A; FRS2; Frs2a; FRS2alpha; FRS2-alpha; SNT; SNT1; SNT-1; SNT2; suc1-associated neurotrophic factor target 1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10818, 314850, 327826 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction