missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Frizzled-5 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 7 publications
£168.00 - £461.00
Specifications
| Antigen | Frizzled-5 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18750678
![]() |
R&D Systems
AF1617-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18032632
![]() |
R&D Systems
AF1617 |
100 μg |
£461.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Frizzled-5 Polyclonal specifically detects Frizzled-5 in Human samples. It is validated for Western Blot.Specifications
| Frizzled-5 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| C2orf31, chromosome 2 open reading frame 31, DKFZP434E2135, frizzled (Drosophila) homolog 5, frizzled homolog 5 (Drosophila), frizzled-5, Frizzled5, Frz-5, FZ-5, FZD5, FzE5, HFZ5, MGC129692, seven-transmembrane receptor frizzled-5, Wnt receptor | |
| FZD5 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| AAC50385 | |
| 7855 | |
| Mouse myeloma cell line NS0-derived recombinant human Frizzled-5 Ala27-Pro167 Accession # AAC50385 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Frizzled-5 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse (rm) Frizzled-1, rmFrizzled-2, rmFrizzled-3, rmFrizzled-6, rmFrizzled-7, and rmFrizzled-8 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title