missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Frizzled-5 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 7 publications
Brand: R&D Systems AF1617-SP
This item is not returnable.
View return policy
Description
Frizzled-5 Polyclonal specifically detects Frizzled-5 in Human samples. It is validated for Western Blot.
Specifications
| Frizzled-5 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C2orf31, chromosome 2 open reading frame 31, DKFZP434E2135, frizzled (Drosophila) homolog 5, frizzled homolog 5 (Drosophila), frizzled-5, Frizzled5, Frz-5, FZ-5, FZD5, FzE5, HFZ5, MGC129692, seven-transmembrane receptor frizzled-5, Wnt receptor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7855 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| AAC50385 | |
| FZD5 | |
| Mouse myeloma cell line NS0-derived recombinant human Frizzled-5 Ala27-Pro167 Accession # AAC50385 | |
| 25 μg | |
| Primary | |
| Detects human Frizzled-5 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse (rm) Frizzled-1, rmFrizzled-2, rmFrizzled-3, rmFrizzled-6, rmFrizzled-7, and rmFrizzled-8 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction