missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frizzled-10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£177.00 - £319.00
Specifications
| Antigen | Frizzled-10 |
|---|---|
| Dilution | Western Blot 1:500-1: 1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18168731
|
Novus Biologicals
NBP2-23659SS |
0.025 mL |
£177.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18107301
|
Novus Biologicals
NBP2-23659 |
0.1 mL |
£319.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Frizzled-10 Polyclonal specifically detects Frizzled-10 in Human, Mouse, Primate samples. It is validated for Western Blot.Specifications
| Frizzled-10 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| GPCR, Growth and Development, Neuronal Cell Markers, Signal Transduction, Wnt Signaling Pathway | |
| 0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide | |
| 11211 | |
| A portion of amino acid 150-200 of human Frizzled-10 was used as the immunogen | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1: 1000 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Primate | |
| CD350, CD350 antigen, frizzled (Drosophila) homolog 10, frizzled homolog 10 (Drosophila), frizzled-10, Fz10, Fz-10, FzE7, hFz10 | |
| FZD10 | |
| IgG | |
| Unpurified | |
| The amino acid sequence used as immunogen is 100% homologous in human, mouse and monkey and 88% homologous in rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title