missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frizzled-10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-23659
This item is not returnable.
View return policy
Description
Frizzled-10 Polyclonal specifically detects Frizzled-10 in Human, Mouse, Primate samples. It is validated for Western Blot.
Specifications
| Frizzled-10 | |
| Polyclonal | |
| Western Blot 1:500-1: 1000 | |
| CD350, CD350 antigen, frizzled (Drosophila) homolog 10, frizzled homolog 10 (Drosophila), frizzled-10, Fz10, Fz-10, FzE7, hFz10 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, mouse and monkey and 88% homologous in rat. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide | |
| FZD10 | |
| A portion of amino acid 150-200 of human Frizzled-10 was used as the immunogen | |
| 0.1 mL | |
| GPCR, Growth and Development, Neuronal Cell Markers, Signal Transduction, Wnt Signaling Pathway | |
| 11211 | |
| Human, Mouse, Primate | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction