Learn More
SPEX™ Freezer/Mill™ High Capacity Cryogenic Grinder
Product Code. 15850447 Shop All SPEX Certiprep Products
Change view
Click to view available options
Voltage:
115/230 V
Unit Size:
Each
Product Code. Voltage unitSize
15850447 115/230 V Each
1 options
Product Code. 15850447 Supplier SPEX™ Supplier No. 6875A230
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
SPEX™ Freezer/Mill™ High Capacity Cryogenic Grinder features a self-contained liquid nitrogen tub and insulated case.
  • Optional auto-fill system is available to automatically fill the bath with liquid nitrogen
  • It has one dual purpose chamber that precools and grinds samples from 0.1 to 100 grams
  • Its grinding chamber holds one large grinding vial, one mid-sized vial, four small grinding vials, or four microvial sets
  • Liquid nitrogen required for operation. Equipped with liquid nitrogen level sensor, insulated case and lid safety interlock switch for operator protection
  • Cryogenic grinding Vial Sets offered in a variety of materials including polycarbonate, stainless steel, and Cr-free steel
  • Grinds samples in the 0.1 - 100 gram range. Cryogenic grinding chamber holds one large grinding vial, one mid-size vial, four standard grinding vials or four microvial sets
  • Auto-Fill System available for automated filling with liquid nitrogen
  • Liquid nitrogen is required for operation
  • The sample’s temperature is maintained because it never leaves the enclosed liquid nitrogen bath
  • Closed cryogenic grinding vial with impactor prevents sample cross-contamination grinding vials made of alternative materials are also available
  • Safety features include LN sensor and lid interlock. If lid is opened during the grinding process grinding will stop
Specifications
Certifications/Compliance CE Approved
Dimensions (L x W x H) 52 x 55 x 46 cm
Display Type Touch Screen
Electrical Requirements 115/230 V, 50/60 Hz
Includes Auto-Fill System
Plug Type 3-prong grounded plug or EU plug
Voltage 115/230 V