missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FPRL1/FPR2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 21 publication
£214.00 - £470.00
Specifications
| Antigen | FPRL1/FPR2 |
|---|---|
| Concentration | 1.1 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry, Immunohistochemistry 1:300, Immunocytochemistry/Immunofluorescence 1:20-1:75, Immunohistochemistry-Paraffin 1:300, Immunohistochemistry-Frozen reported in scientific literature (PMID 24086560), Electron Microscopy |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18709773
|
Novus Biologicals
NLS1878SS |
0.025 mL |
£214.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18797423
|
Novus Biologicals
NLS1878 |
£470.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
FPRL1/FPR2 Polyclonal specifically detects FPRL1/FPR2 in Human, Mouse, Rat, Bacteria samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy.Specifications
| FPRL1/FPR2 | |
| Western Blot 1:1000, Flow Cytometry, Immunohistochemistry 1:300, Immunocytochemistry/Immunofluorescence 1:20-1:75, Immunohistochemistry-Paraffin 1:300, Immunohistochemistry-Frozen reported in scientific literature (PMID 24086560), Electron Microscopy | |
| Polyclonal | |
| Rabbit | |
| Immunology | |
| PBS, 30% Glycerol with 0.01% Sodium Azide | |
| ALXR, FMLP-R-II, FMLPX, formyl peptide receptor 2, Formyl peptide receptor-like 1RFP, FPR2A, FPRH1FMLP-related receptor I, FPRL1LXA4 receptor, HM63FPRH2, Lipoxin A4 receptor, lipoxin A4 receptor (formyl peptide receptor related), LXA4RFMLP-R-I, N-formyl peptide receptor 2 | |
| FPR2 | |
| IgG | |
| Affinity Purified | |
| Human and mouse. Rat reactivity reported in scientific literature (PMID: 24086560). |
| 1.1 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Bacteria | |
| P25090 | |
| 2358 | |
| This FPRL1/FPR2 antibody is made to a synthetic peptide made to the 2nd extracellular loop of the human FPRL1 protein (between residues 300-350) [UniProt P25090] | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| 38 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title