missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FPRL1/FPR2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 21 publication
Brand: Novus Biologicals NLS1878SS
This item is not returnable.
View return policy
Description
FPRL1/FPR2 Polyclonal specifically detects FPRL1/FPR2 in Human, Mouse, Rat, Bacteria samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy.
Specifications
| FPRL1/FPR2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 30% Glycerol with 0.01% Sodium Azide | |
| ALXR, FMLP-R-II, FMLPX, formyl peptide receptor 2, Formyl peptide receptor-like 1RFP, FPR2A, FPRH1FMLP-related receptor I, FPRL1LXA4 receptor, HM63FPRH2, Lipoxin A4 receptor, lipoxin A4 receptor (formyl peptide receptor related), LXA4RFMLP-R-I, N-formyl peptide receptor 2 | |
| Rabbit | |
| 38 kDa | |
| 0.025 mL | |
| Immunology | |
| 2358 | |
| Human, Mouse, Rat, Bacteria | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay | |
| 1.1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry, Immunohistochemistry 1:300, Immunocytochemistry/Immunofluorescence 1:20-1:75, Immunohistochemistry-Paraffin 1:300, Immunohistochemistry-Frozen reported in scientific literature (PMID 24086560), Electron Microscopy | |
| P25090 | |
| FPR2 | |
| This FPRL1/FPR2 antibody is made to a synthetic peptide made to the 2nd extracellular loop of the human FPRL1 protein (between residues 300-350) [UniProt P25090] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and mouse. Rat reactivity reported in scientific literature (PMID: 24086560). | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction