missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FPGS Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
£161.00 - £383.00
Specifications
| Antigen | FPGS |
|---|---|
| Dilution | Western Blot 1:100 - 1:500, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232310
|
Novus Biologicals
NBP3-33257-20ul |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30230012
|
Novus Biologicals
NBP3-33257-100ul |
100 μL |
£383.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FPGS Monoclonal antibody specifically detects FPGS in Human,Rat samples. It is validated for ELISA,Western BlotSpecifications
| FPGS | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Rat | |
| EC 6.3.2.17, Folylpoly-gamma-glutamate synthetase, folylpolyglutamate synthase, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 450-587 of human FPGS (Q05932).,, Sequence:, NADQQNFTVTLDQVLLRCLEHQQHWNHLDEEQASPDLWSAPSPEPGGSASLLLAPHPPHTCSASSLVFSCISHALQWISQGRDPIFQPPSPPKGLLTHPVAHSGASILREAAAIHVLVTGSLHLVGGVLKLLEPALSQ | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:100 - 1:500, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 2356 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title