missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FPGS Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33257-20ul
This item is not returnable.
View return policy
Description
FPGS Monoclonal antibody specifically detects FPGS in Human,Rat samples. It is validated for ELISA,Western Blot
Specifications
| FPGS | |
| Monoclonal | |
| Western Blot 1:100 - 1:500, ELISA Recommended starting concentration is 1 μg/mL | |
| EC 6.3.2.17, Folylpoly-gamma-glutamate synthetase, folylpolyglutamate synthase, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 450-587 of human FPGS (Q05932).,, Sequence:, NADQQNFTVTLDQVLLRCLEHQQHWNHLDEEQASPDLWSAPSPEPGGSASLLLAPHPPHTCSASSLVFSCISHALQWISQGRDPIFQPPSPPKGLLTHPVAHSGASILREAAAIHVLVTGSLHLVGGVLKLLEPALSQ | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 2356 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction