missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXO3, p Ser253 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£219.00 - £341.00
Specifications
| Antigen | FOXO3 (p Ser253) |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18280104
|
Novus Biologicals
NB100-81929-0.025ML |
0.025 mL |
£219.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18084574
|
Novus Biologicals
NB100-81929 |
0.05 mL |
£341.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FOXO3 Polyclonal specifically detects FOXO3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| FOXO3 (p Ser253) | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, DNA Repair, Phospho Specific, Transcription Factors and Regulators | |
| O43524 | |
| 2309 | |
| IgG | |
| Affinity Purified | |
| 97 kDa |
| 1.0 mg/mL | |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Human, Rat, Mouse | |
| AF6q21, AF6q21 protein, DKFZp781A0677, FKHRL1MGC31925, FKHRL1P2, forkhead box O3, forkhead box protein O3, forkhead homolog (rhabdomyosarcoma) like 1, Forkhead in rhabdomyosarcoma-like 1, forkhead, Drosophila, homolog of, in rhabdomyosarcoma-like 1, FOXO2, FOXO3Aforkhead box O3A, MGC12739 | |
| FOXO3 | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title