missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXO3, p Ser253 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-81929
This item is not returnable.
View return policy
Description
FOXO3 Polyclonal specifically detects FOXO3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| FOXO3 (p Ser253) | |
| Polyclonal | |
| Unconjugated | |
| PBS (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, 50% glycerol with 0.02% Sodium Azide | |
| AF6q21, AF6q21 protein, DKFZp781A0677, FKHRL1MGC31925, FKHRL1P2, forkhead box O3, forkhead box protein O3, forkhead homolog (rhabdomyosarcoma) like 1, Forkhead in rhabdomyosarcoma-like 1, forkhead, Drosophila, homolog of, in rhabdomyosarcoma-like 1, FOXO2, FOXO3Aforkhead box O3A, MGC12739 | |
| Rabbit | |
| 97 kDa | |
| 0.05 mL | |
| Apoptosis, Cancer, DNA Repair, Phospho Specific, Transcription Factors and Regulators | |
| 2309 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100 | |
| O43524 | |
| FOXO3 | |
| The antiserum was produced against synthesized phosphopeptide derived from human FOXO3 around the phosphorylation site of serine 253 (A-V-SP-M-D). | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Detects endogenous levels of FOXO3 only when phosphorylated at serine 253. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction