Learn More
Guest Medical™ Four Tier Mobile Laboratory Trolley
In many areas of health care there is a need to have instruments, dressings and equipment readily to hand in a number of different working locations. The versatile trolley has been designed to fulfill this requirement, and has been found to be ideally suited for use in areas such as laboratories, wards, clinics, theatres, phlebotomy units and chiropody departments.
Brand:  Guest Medical™ S9618
Additional Details : Weight : 13.00000kg
 View more versions of this product
Product Code. 16337535
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Designed for those staff that usually stand whilst working
  • Pivoting drawers for total accessibility
  • Choice of size and drawer options to suit individual user needs
  • Manufactured from high quality ABS for strength and durability
  • Mounted on five castors ensuring extra stability and manoeuverability
  • Easily cleaned using wipes or spray
Specifications
Specifications
White
5
13.0 kg
Drawers: 60 mm
6