Learn More
Watson-Marlow 314DW Pumphead
Product Code. 12476340
Change view
Click to view available options
For Use With:
323 Series Peristaltic Pumps
Unit Size:
Each
Product Code. For Use With Unit Size
12476340 323 Series Peristaltic Pumps Each
1 options
Product Code. 12476340 Supplier Watson-Marlow Supplier No. 033.4451.000
  • ONLINE EXCLUSIVE PRICE
    £720.35
    £333.04 / Pack of 1
    Save £387.31
    53% Off
View all promotions
Estimated Shipment: 19-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
300 Series Pumpheads. Flip top pumpheads for continuous tubing for flow rates to 3 L/min. Extendable up to 6 channels of flow.
  • The 300 series combines performance, practicality and design. With its iconic flip-top design it is an industry standard OEM product in medical device, biotechnology and diagnostic applications, and very popular in many other applications too.
  • Case material: Glass filled polypropylene, IXEF (Polyarylamide), and Stainless steel
  • Rotor material: Electroless nickel plated hardened steel, and Glass filled Nylon
TRUSTED_SUSTAINABILITY
Specifications
For Use With 323 Series Peristaltic Pumps
No. of Rollers 4
Type Pump Head