Learn More
Watson-Marlow 314X Pumphead
Product Code. 12496340
Change view
Click to view available options
For Use With:
323 Series Peristaltic Pumps
Unit Size:
Each
Product Code. For Use With Unit Size
12496340 323 Series Peristaltic Pumps Each
1 options
Product Code. 12496340 Supplier Watson-Marlow Supplier No. 033.4431.000
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
300 Series Pumpheads. Flip top pumpheads for continuous tubing for flow rates to 3 L/min. Extendable up to 6 channels of flow.
  • The 300 series combines performance, practicality and design. With its iconic flip-top design it is an industry standard OEM product in medical device, biotechnology and diagnostic applications, and very popular in many other applications too.
  • Case material: Glass filled polypropylene, IXEF (Polyarylamide), and Stainless steel
  • Rotor material: Electroless nickel plated hardened steel, and Glass filled Nylon
TRUSTED_SUSTAINABILITY
Specifications
For Use With 323 Series Peristaltic Pumps
Material Polypropylene
No. of Rollers 4
Type Pump Extension
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.