missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOS Mouse anti-Human, Mouse, Clone: 1G2C5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66590-1-IG-150UL
This item is not returnable.
View return policy
Description
FBJ murine osteosarcoma viral oncogene homolog is commonly known as Cellular protooncogene Fos (c-Fos). It belongs to the fos transcription factor family and is encoded by the C-FOS gene located on chromosome 14. c-Fos heterodimerizes with c-Jun to form the Activator Protein -1 (AP-1) transcription factor which regulates transcription of several genes that play an important role in cellular signal transduction, cell proliferation and differentiation. c-Fos has been shown to be phosphorylated by various stimuli including growth factors and insulin at atleast seven different sites. The best known upstream kinase for phosphorylation at Threonine 232 is ERK MAPK which regulates localization of c-Fos to the nucleus and is thus important for c-Fos induced transcriptional activity.Specifications
| FOS | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P01100, P01101 | |
| FOS | |
| FOS Fusion Protein Ag24340 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1G2C5 | |
| Unconjugated | |
| FOS | |
| AP 1, C FOS, Cellular oncogene fos, FOS, G0S7, Proto oncogene c Fos | |
| Mouse | |
| Protein G | |
| RUO | |
| 14281, 2353 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction